5-Bromo-2-cyanonicotinic acid 90%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-3-carboxypicolinonitrile, 5-Bromo-2-cyanopyridine-3-carboxylic acid, 5-Bromo-3-carboxy-2-cyanopyridine
OR8997-500MG
OR8997-100MG
APOSOR8997-500MGEA
781
EUR
APOSOR8997-500MG
APOSOR8997-100MG
5-Bromo-2-cyanonicotinic acid 90%
5-Bromo-2-cyanonicotinic acid
Formel:
C₇H₃BrN₂O₂ Molecular Weight: 227,015 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08543463 CAS-Nummer: 914637-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...