4-(Allyloxy)benzonitrile 95%
Lieferant: Apollo Scientific
OR900067-1G
OR900067-5G
APOSOR900067-1GEA
236
EUR
APOSOR900067-1G
APOSOR900067-5G
4-(Allyloxy)benzonitrile 95%
4-(Allyloxy)benzonitrile
Formel:
C₁₀H₉NO Molecular Weight: 159,185 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09940590 CAS-Nummer: 33148-47-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...