1-Bromo-4-(cyclopropylmethoxy)benzene 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR900180-1G
OR900180-5G
APOSOR900180-1GEA
227
EUR
APOSOR900180-1G
APOSOR900180-5G
1-Bromo-4-(cyclopropylmethoxy)benzene 98%
1-Bromo-4-(cyclopropylmethoxy)benzene
Formel:
C₁₀H₁₁BrO Molecular Weight: 227,098 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12138829 CAS-Nummer: 412004-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...