1-Bromo-2-chloro-4-pyrrolidinobenzene 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromo-3-chlorophenyl)pyrrolidine

OR900184-1G OR900184-5G
APOSOR900184-1GEA 114 EUR
APOSOR900184-1G APOSOR900184-5G
1-Bromo-2-chloro-4-pyrrolidinobenzene 98%
1-Bromo-2-chloro-4-pyrrolidinobenzene
Formel: C₁₀H₁₁BrClN
Molecular Weight: 260,558 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18837620
CAS-Nummer: 1291487-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR