3-(Ethanesulphonyl)aniline 96%

Lieferant: Apollo Scientific

Synonyme: 3-(Ethylsulphonyl)aniline

OR900357-1G OR900357-250MG
APOSOR900357-1GEA 580 EUR
APOSOR900357-1G APOSOR900357-250MG
3-(Ethanesulphonyl)aniline 96%
3-(Ethanesulphonyl)aniline
Formel: C₈H₁₁NO₂S
Molecular Weight: 185,243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11189664
CAS-Nummer: 83164-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR