N,N'-Bis(tert-butoxycarbonyl)-o-phenylenediamine 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl N-(2-{[(tert-butoxy)carbonyl]amino}phenyl)carbamate, 1,2-Diaminobenzene, 1,2-BOC protected

OR900427-1G OR900427-5G
APOSOR900427-1GEA 147 EUR
APOSOR900427-1G APOSOR900427-5G
N,N'-Bis(tert-butoxycarbonyl)-o-phenylenediamine 96%
N,N'-Bis(tert-butoxycarbonyl)-o-phenylenediamine
Formel: C₁₆H₂₄N₂O₄
Molecular Weight: 308,373 g/mol
Schmelzpunkt: 104…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD24465598
CAS-Nummer: 438533-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR