6-Methoxy-1-benzothiophene 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methoxybenzo[b]thiophene

OR900495-1G OR900495-250MG
APOSOR900495-1GEA 372 EUR
APOSOR900495-1G APOSOR900495-250MG
6-Methoxy-1-benzothiophene 95%
6-Methoxy-1-benzothiophene
Formel: C₉H₈OS
Molecular Weight: 164.224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13181215
CAS-Nummer: 90560-10-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR