6-Methoxy-1-benzofuran 96%

Lieferant: Apollo Scientific

Synonyme: 6-Methoxybenzofuran

OR900496-1G OR900496-250MG
APOSOR900496-1GEA 112 EUR
APOSOR900496-1G APOSOR900496-250MG
6-Methoxy-1-benzofuran 96%
6-Methoxy-1-benzofuran
Formel: C₉H₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11656393
CAS-Nummer: 50551-63-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR