1-Chloro-3-(ethoxymethyl)benzene 96%
Lieferant: Apollo Scientific
OR900531-1G
OR900531-5G
APOSOR900531-1GEA
154
EUR
APOSOR900531-1G
APOSOR900531-5G
1-Chloro-3-(ethoxymethyl)benzene 96%
1-Chloro-3-(ethoxymethyl)benzene
Formel:
C₉H₁₁ClO Molecular Weight: 170.636 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23699511 CAS-Nummer: 68578-50-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...