1-Cyclopentylpyrazol-4-amine 95%
Lieferant: Apollo Scientific
OR900558-1G
OR900558-250MG
APOSOR900558-1GEA
877
EUR
APOSOR900558-1G
APOSOR900558-250MG
1-Cyclopentylpyrazol-4-amine 95%
1-Cyclopentylpyrazol-4-amine
Formel:
C₈H₁₃N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11128189 CAS-Nummer: 1152866-89-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...