tert-Butyl 2-(2-oxopyrrolidin-1-yl)acetate 98%

Lieferant: Apollo Scientific

OR900571-1G OR900571-5G
APOSOR900571-1GEA 219 EUR
APOSOR900571-1G APOSOR900571-5G
tert-Butyl 2-(2-oxopyrrolidin-1-yl)acetate 98%
tert-Butyl 2-(2-oxopyrrolidin-1-yl)acetate
Formel: C₁₀H₁₇NO₃
Molecular Weight: 199,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23844302
CAS-Nummer: 497250-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR