4-Aminooxane-4-carboxylic acid hydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Aminotetrahydro-2H-pyran-4-carboxylic acid hydrochloride
OR900599-1G
OR900599-5G
APOSOR900599-1GEA
40.6
EUR
APOSOR900599-1G
APOSOR900599-5G
4-Aminooxane-4-carboxylic acid hydrochloride 95%
4-Aminooxane-4-carboxylic acid hydrochloride
Formel:
C₆H₁₁NO₃·ClH Molecular Weight: 181.617 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683135 CAS-Nummer: 217299-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...