N-(4-Methoxybenzyl)maleimide 95%
Lieferant: Apollo Scientific
Synonyme:
1-[(4-Methoxyphenyl)methyl]pyrrole-2,5-dione
OR900618-10G
OR900618-1G
OR900618-5G
APOSOR900618-10GEA
1140
EUR
APOSOR900618-10G
APOSOR900618-1G
APOSOR900618-5G
N-(4-Methoxybenzyl)maleimide 95%
N-(4-Methoxybenzyl)maleimide
Formel:
C₁₂H₁₁NO₃ Molecular Weight: 217,221 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00113970 CAS-Nummer: 140480-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...