(1-Methylcyclohexyl)methanol 97%

Lieferant: Apollo Scientific

OR900633-1G OR900633-250MG
APOSOR900633-1GEA 277 EUR
APOSOR900633-1G APOSOR900633-250MG
(1-Methylcyclohexyl)methanol 97%
(1-Methylcyclohexyl)methanol
Formel: C₈H₁₆O
Molecular Weight: 128,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00270318
CAS-Nummer: 14064-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR