N-Cyclopropyl-3-hydroxybenzamide 98%
Lieferant: Apollo Scientific
OR900651-1G
OR900651-250MG
OR900651-5G
APOSOR900651-1GEA
270
EUR
APOSOR900651-1G
APOSOR900651-250MG
APOSOR900651-5G
N-Cyclopropyl-3-hydroxybenzamide 98%
N-Cyclopropyl-3-hydroxybenzamide
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177.2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11130687 CAS-Nummer: 1019466-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...