4-(Pyrrolidinocarbonyl)phenol 98%

Lieferant: Apollo Scientific

OR900668-1G OR900668-5G
APOSOR900668-1GEA 0 EUR
APOSOR900668-1G APOSOR900668-5G
4-(Pyrrolidinocarbonyl)phenol 98%
4-(Pyrrolidinocarbonyl)phenol
Formel: C₁₁H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07633880
CAS-Nummer: 478929-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR