1-Phenylcyclopropane-1-carboxamide 96%

Lieferant: Apollo Scientific

OR900735-1G OR900735-5G
APOSOR900735-1GEA 219 EUR
APOSOR900735-1G APOSOR900735-5G
1-Phenylcyclopropane-1-carboxamide 96%
1-Phenylcyclopropane-1-carboxamide
Formel: C₁₀H₁₁NO
Molecular Weight: 161.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00463735
CAS-Nummer: 6120-96-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR