2-Acetamido-3,3-dimethylbutanoic acid 96%
Lieferant: Apollo Scientific
OR900748-1G
OR900748-5G
APOSOR900748-1GEA
270
EUR
APOSOR900748-1G
APOSOR900748-5G
2-Acetamido-3,3-dimethylbutanoic acid 96%
2-Acetamido-3,3-dimethylbutanoic acid
Formel:
C₈H₁₅NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08691200 CAS-Nummer: 22146-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...