(2-Chloro-5-methylphenyl)methanol 97%
Lieferant: Apollo Scientific
OR900762-1G
OR900762-250MG
APOSOR900762-1GEA
632
EUR
APOSOR900762-1G
APOSOR900762-250MG
(2-Chloro-5-methylphenyl)methanol 97%
(2-Chloro-5-methylphenyl)methanol
Formel:
C₈H₉ClO Molecular Weight: 156,609 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14155858 CAS-Nummer: 89981-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...