2-Chloro-3-methylbenzyl alcohol 98%

Lieferant: Apollo Scientific

Synonyme: (2-Chloro-3-methylphenyl)methanol

OR900817-1G OR900817-250MG
APOSOR900817-1GEA 290 EUR
APOSOR900817-1G APOSOR900817-250MG
2-Chloro-3-methylbenzyl alcohol 98%
2-Chloro-3-methylbenzyl alcohol
Formel: C₈H₉ClO
Molecular Weight: 156,609 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12031743
CAS-Nummer: 61563-27-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR