4-Thiophen-2-ylphenol 98%

Lieferant: Apollo Scientific

Synonyme: 4-(Thiophen-2-yl)phenol

OR900821-1G
APOSOR900821-1GEA 161 EUR
APOSOR900821-1G
4-Thiophen-2-ylphenol 98%
4-Thiophen-2-ylphenol
Formel: C₁₀H₈OS
Molecular Weight: 176,235 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797407
CAS-Nummer: 29886-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR