5,5'-Dimethyldipyrromethane 95%

Lieferant: Apollo Scientific

OR900845-1G OR900845-500MG
APOSOR900845-1GEA 684 EUR
APOSOR900845-1G APOSOR900845-500MG
5,5'-Dimethyldipyrromethane 95%
5,5'-Dimethyldipyrromethane
Formel: C₁₁H₁₄N₂
Molecular Weight: 174,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669617
CAS-Nummer: 99840-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR