3-Methoxyisoxazole-5-carboxylic acid 95%

Lieferant: Apollo Scientific

OR900847-1G OR900847-500MG
APOSOR900847-1GEA 400 EUR
APOSOR900847-1G APOSOR900847-500MG
3-Methoxyisoxazole-5-carboxylic acid 95%
3-Methoxyisoxazole-5-carboxylic acid
Formel: C₅H₅NO₄
Molecular Weight: 143.098 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368245
CAS-Nummer: 13626-59-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR