1-(3-Bromophenyl)cyclopropanamine 95%

Lieferant: Apollo Scientific
Achtung

OR900848-10G OR900848-1G OR900848-5G
APOSOR900848-10GEA 920 EUR
APOSOR900848-10G APOSOR900848-1G APOSOR900848-5G
1-(3-Bromophenyl)cyclopropanamine 95%
1-(3-Bromophenyl)cyclopropanamine
Formel: C₉H₁₀BrN
Molecular Weight: 212,086 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374476
CAS-Nummer: 546115-65-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR