1,5-Naphthyridin-2(1H)-one 98%
Lieferant: Apollo Scientific
OR901584-1G
APOSOR901584-1GEA
59.6
EUR
APOSOR901584-1G
1,5-Naphthyridin-2(1H)-one 98%
1,5-Naphthyridin-2(1H)-one
Formel:
C₈H₆N₂O Molecular Weight: 146,146 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11877925 CAS-Nummer: 10261-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...