2,4-Dimethoxypyridine 98%

Lieferant: Apollo Scientific

OR901626-25G
APOSOR901626-25GEA 150 EUR
APOSOR901626-25G
2,4-Dimethoxypyridine 98%
2,4-Dimethoxypyridine
Formel: C₇H₉NO₂
Molecular Weight: 139,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01646187
CAS-Nummer: 18677-43-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR