8-Chloroquinolin-2-ol 98%

Lieferant: Apollo Scientific

OR901653-100G OR901653-25G
APOSOR901653-100GEA 328 EUR
APOSOR901653-100G APOSOR901653-25G
8-Chloroquinolin-2-ol 98%
8-Chloroquinolin-2-ol
Formel: C₉H₆ClNO
Molecular Weight: 179,603 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03427166
CAS-Nummer: 23981-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR