2-Ethoxyquinoline 98%

Lieferant: Apollo Scientific

OR901677-1G OR901677-25G OR901677-5G
APOSOR901677-1GEA 145 EUR
APOSOR901677-1G APOSOR901677-25G APOSOR901677-5G
2-Ethoxyquinoline 98%
2-Ethoxyquinoline
Formel: C₁₁H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00234738
CAS-Nummer: 46185-83-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR