2-Methoxy-6-phenylpyridine 95%
Lieferant: Apollo Scientific
OR901685-1G
OR901685-5G
APOSOR901685-1GEA
145
EUR
APOSOR901685-1G
APOSOR901685-5G
2-Methoxy-6-phenylpyridine 95%
2-Methoxy-6-phenylpyridine
Formel:
C₁₂H₁₁NO Molecular Weight: 185.222 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044374 CAS-Nummer: 35070-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...