6,8-Dimethylquinoline hydrochloride 96%

Lieferant: Apollo Scientific

OR901724-100G OR901724-25G OR901724-5G
APOSOR901724-100GEA 632 EUR
APOSOR901724-100G APOSOR901724-25G APOSOR901724-5G
6,8-Dimethylquinoline hydrochloride 96%
6,8-Dimethylquinoline hydrochloride
Formel: C₁₁H₁₁N·ClH
Molecular Weight: 193,673 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17214294
CAS-Nummer: 1255574-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR