2-Chloro-4-piperidinopyridine 98%
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-4-(piperidin-1-yl)pyridine
OR901746-1G
OR901746-5G
APOSOR901746-1GEA
168
EUR
APOSOR901746-1G
APOSOR901746-5G
2-Chloro-4-piperidinopyridine 98%
2-Chloro-4-piperidinopyridine
Formel:
C₁₀H₁₃ClN₂ Molecular Weight: 196.677 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14702602 CAS-Nummer: 1209459-54-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...