3-bromo-N-methylpyridin-2-amine 98%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-N-methylpyridin-2-amine

OR901758-1G OR901758-5G
APOSOR901758-1GEA 79.9 EUR
APOSOR901758-1G APOSOR901758-5G
3-bromo-N-methylpyridin-2-amine 98%
3-bromo-N-methylpyridin-2-amine
Formel: C₆H₇BrN₂
Molecular Weight: 187.037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374349
CAS-Nummer: 214977-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR