3,5,6-Trichloropicoline 98%

Lieferant: Apollo Scientific

OR901783-1G OR901783-5G
APOSOR901783-1GEA 145 EUR
APOSOR901783-1G APOSOR901783-5G
3,5,6-Trichloropicoline 98%
3,5,6-Trichloropicoline
Formel: C₆H₄Cl₃N
Molecular Weight: 196,462 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18783195
CAS-Nummer: 22109-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR