2-(6-bromopyridin-2-yl)acetonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-6-(cyanomethyl)pyridine

OR901849-1G OR901849-250MG
APOSOR901849-1GEA 195 EUR
APOSOR901849-1G APOSOR901849-250MG
2-(6-bromopyridin-2-yl)acetonitrile 95%
2-(6-bromopyridin-2-yl)acetonitrile
Formel: C₇H₅BrN₂
Molecular Weight: 197,032 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08706174
CAS-Nummer: 112575-11-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR