2-Phenylpyridine-3-carbonitrile 96%

Lieferant: Apollo Scientific

OR901868-1G OR901868-5G
APOSOR901868-1GEA 168 EUR
APOSOR901868-1G APOSOR901868-5G
2-Phenylpyridine-3-carbonitrile 96%
2-Phenylpyridine-3-carbonitrile
Formel: C₁₂H₈N₂
Molecular Weight: 180.205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02929371
CAS-Nummer: 39065-49-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR