3-Methoxy-5-phenylpyridine 98%

Lieferant: Apollo Scientific

OR901878-1G OR901878-250MG
APOSOR901878-1GEA 364 EUR
APOSOR901878-1G APOSOR901878-250MG
3-Methoxy-5-phenylpyridine 98%
3-Methoxy-5-phenylpyridine
Formel: C₁₂H₁₁NO
Molecular Weight: 185,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14702089
CAS-Nummer: 53698-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR