3-Chloro-4-ethoxyaniline 98%
Lieferant: Apollo Scientific
OR902121-10G
OR902121-1G
OR902121-5G
APOSOR902121-10GEA
285
EUR
APOSOR902121-10G
APOSOR902121-1G
APOSOR902121-5G
3-Chloro-4-ethoxyaniline 98%
3-Chloro-4-ethoxyaniline
Formel:
C₈H₁₀ClNO Molecular Weight: 171.624 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08687289 CAS-Nummer: 5211-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...