2-chloro-5,6,7,8-tetrahydroquinoline 95%
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-5,6,7,8-tetrahydroquinoline
OR902126-1G
OR902126-250MG
APOSOR902126-1GEA
242
EUR
APOSOR902126-1G
APOSOR902126-250MG
2-chloro-5,6,7,8-tetrahydroquinoline 95%
2-chloro-5,6,7,8-tetrahydroquinoline
Formel:
C₉H₁₀ClN Molecular Weight: 167,635 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09034992 CAS-Nummer: 21172-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...