2-Phenylazepane 96%

Lieferant: Apollo Scientific

OR902176-1G OR902176-5G
APOSOR902176-1GEA 342 EUR
APOSOR902176-1G APOSOR902176-5G
2-Phenylazepane 96%
2-Phenylazepane
Formel: C₁₂H₁₇N
Lagertemperatur: Raumtemperatur
MDL: MFCD02663698
CAS-Nummer: 3466-82-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR