1-Methyl-1H-indol-4-ol 95%

Lieferant: Apollo Scientific

OR902258-1G OR902258-250MG
APOSOR902258-1GEA 155 EUR
APOSOR902258-1G APOSOR902258-250MG
1-Methyl-1H-indol-4-ol 95%
1-Methyl-1H-indol-4-ol
Formel: C₉H₉NO
Molecular Weight: 147.174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834885
CAS-Nummer: 7556-37-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR