N,N-Dimethylpivalamide 98%

Lieferant: Apollo Scientific

OR902440-10G OR902440-1G OR902440-5G
APOSOR902440-10GEA 899 EUR
APOSOR902440-10G APOSOR902440-1G APOSOR902440-5G
N,N-Dimethylpivalamide 98%
N,N-Dimethylpivalamide
Formel: C₇H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00043644
CAS-Nummer: 24331-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR