2-(2-Methylpropyl)azepane 95%
Lieferant: Apollo Scientific
OR902479-1G
OR902479-250MG
APOSOR902479-1GEA
806
EUR
APOSOR902479-1G
APOSOR902479-250MG
2-(2-Methylpropyl)azepane 95%
2-(2-Methylpropyl)azepane
Formel:
C₁₀H₂₁N Molecular Weight: 155,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02663711 CAS-Nummer: 80053-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...