4-Chloroquinolin-6-amine 97%

Lieferant: Apollo Scientific

OR902593-1G OR902593-250MG
APOSOR902593-1GEA 517 EUR
APOSOR902593-1G APOSOR902593-250MG
4-Chloroquinolin-6-amine 97%
4-Chloroquinolin-6-amine
Formel: C₉H₇ClN₂
Molecular Weight: 178.618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14702408
CAS-Nummer: 1085192-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR