N-(4-Hydroxyphenyl)maleimide 98%
Lieferant: Apollo Scientific
Synonyme:
1-(4-Hydroxyphenyl)-1H-pyrrole-2,5-dione
OR902636-100G
APOSOR902636-100GEA
148
EUR
APOSOR902636-100G
N-(4-Hydroxyphenyl)maleimide 98%
N-(4-Hydroxyphenyl)maleimide
Formel:
C₁₀H₇NO₃ Molecular Weight: 189,167 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129167 CAS-Nummer: 7300-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...