6-Ethyl-1H-benzimidazol-2-ylamine 97%

Lieferant: Apollo Scientific

OR902739-1G OR902739-5G
APOSOR902739-1GEA 227 EUR
APOSOR902739-1G APOSOR902739-5G
6-Ethyl-1H-benzimidazol-2-ylamine 97%
6-Ethyl-1H-benzimidazol-2-ylamine
Formel: C₉H₁₁N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04967056
CAS-Nummer: 107726-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR