2-[Methyl(2-nitrophenyl)amino]ethanol 98%

Lieferant: Apollo Scientific

OR902941-1G OR902941-5G
APOSOR902941-1GEA 219 EUR
APOSOR902941-1G APOSOR902941-5G
2-[Methyl(2-nitrophenyl)amino]ethanol 98%
2-[Methyl(2-nitrophenyl)amino]ethanol
Formel: C₉H₁₂N₂O₃
Molecular Weight: 196,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14648506
CAS-Nummer: 106744-74-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR