1-(3-Chloropropyl)pyrrolidin-2-one 95%

Lieferant: Apollo Scientific

OR903411-1G OR903411-250MG
APOSOR903411-1GEA 435 EUR
APOSOR903411-1G APOSOR903411-250MG
1-(3-Chloropropyl)pyrrolidin-2-one 95%
1-(3-Chloropropyl)pyrrolidin-2-one
Formel: C₇H₁₂ClNO
Molecular Weight: 161.629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09742124
CAS-Nummer: 91152-30-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR