2-(2-Thienyl)piperidine 95%

Lieferant: Apollo Scientific

Synonyme: 2-(Thiophen-2-yl)piperidine

OR903976-1G OR903976-250MG
APOSOR903976-1GEA 394 EUR
APOSOR903976-1G APOSOR903976-250MG
2-(2-Thienyl)piperidine 95%
2-(2-Thienyl)piperidine
Formel: C₉H₁₃NS
Molecular Weight: 167.271 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02663548
CAS-Nummer: 270902-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR