1-(3-Methylphenyl)-1H-pyrrole-2-carbaldehyde 95%

Lieferant: Apollo Scientific

OR904018-1G OR904018-5G
APOSOR904018-1GEA 290 EUR
APOSOR904018-1G APOSOR904018-5G
1-(3-Methylphenyl)-1H-pyrrole-2-carbaldehyde 95%
1-(3-Methylphenyl)-1H-pyrrole-2-carbaldehyde
Formel: C₁₂H₁₁NO
Molecular Weight: 185.222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664904
CAS-Nummer: 86454-35-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR