2-Methoxy-5-(2-pyrrolidinyl)pyridine 95%
Lieferant: Apollo Scientific
Synonyme:
2-Methoxy-5-(pyrrolidin-2-yl)pyridine
OR904134-1G
OR904134-250MG
APOSOR904134-1GEA
533
EUR
APOSOR904134-1G
APOSOR904134-250MG
2-Methoxy-5-(2-pyrrolidinyl)pyridine 95%
2-Methoxy-5-(2-pyrrolidinyl)pyridine
Formel:
C₁₀H₁₄N₂O Molecular Weight: 178.231 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05189295 CAS-Nummer: 185510-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...